Learn More
Pyrimidine-2-sulfonyl Fluoride, TRC
Product Code. 30332593
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30332593 50 mg 50mg
1 options
Product Code. 30332593 Supplier Toronto Research Chemicals Supplier No. TRCB52611850MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyrimidine-2-sulfonyl Fluoride
CAS 35762-87-9
Molecular Formula C4H3FN2O2S
Quantity 50 mg
InChI Formula InChI=1S/C4H3FN2O2S/c5-10(8,9)4-6-2-1-3-7-4/h1-3H
SMILES C1=CN=C(N=C1)S(=O)(=O)F
IUPAC Name pyrimidine-2-sulfonyl fluoride
Molecular Weight (g/mol) 162.14
Formula Weight 161.9899