Läs mer
STEMCELL Technologies Pyrintegrin
Beskrivning
- Pyrintegrin is a 2,4-disubstituted pyrimidine that induces the activation of β1 integrin and multiple growth factor receptors, including FGFR1, IGFR1, EGFR1, and HER2 (Xu et al.)
- Enhances integrin-dependent attachment and survival of human embryonic stem (ES) cells following trypsin-mediated single-cell dissociation (Xu et al.)
Specifikationer
Specifikationer
| CAS | 1228445-38-2 |
| Kvantitet | 10 mg |
| Molekylformel | C23H25N5O3S |
| Renhet | ≥95% |