missing translation for 'onlineSavingsMsg'
Läs mer

Pyrithiamine, TRC

Produktkod. 30426662
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30426662 50 mg
30427888 10 mg
30409679 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426662 Supplier Toronto Research Chemicals Supplier No. TRCP99741550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-64-5
Molekylformel C14 H19 N4 O . H Br . Br
Molekylvikt (g/mol) 420.15
Synonym Neopyrithiamine, Pyrithiamin, Pyrithiamin Hydrobromide, Pyrithiamine Bromide Hydrobromide, Pyrithiamine Hydrobromide
IUPAC-namn 1-((4-amino-2-methylpyrimidin-5-yl)methyl)-3-(2-hydroxyethyl)-2-methylpyridin-1-ium bromide hydrobromide
LEDER [Br-].Br.OCCC1=CC=C[N+](=C1C)CC2=CN=C(N=C2N)C

Specifications

Kemiskt namn eller material Pyrithiamine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H19N4O.2BrH/c1-10-12(5-7-19)4-3-6-18(10)9-13-8-16-11(2)17-14(13)15;;/h3-4,6,8,19H,5,7,9H2,1-2H3,(H2,15,16,17);2*1H/q+1;;/p-1
Formel vikt 418.0004
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.