Läs mer
Pyrithiamine, TRC
Produktkod. 30427888
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427888 10 mg
30426662 50 mg
30409679 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99741510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 534-64-5
Molekylformel C14 H19 N4 O . H Br . Br
Molekylvikt (g/mol) 420.15
Synonym Neopyrithiamine, Pyrithiamin, Pyrithiamin Hydrobromide, Pyrithiamine Bromide Hydrobromide, Pyrithiamine Hydrobromide
IUPAC-namn 1-((4-amino-2-methylpyrimidin-5-yl)methyl)-3-(2-hydroxyethyl)-2-methylpyridin-1-ium bromide hydrobromide
LEDER [Br-].Br.OCCC1=CC=C[N+](=C1C)CC2=CN=C(N=C2N)C
Specifikationer
Kemiskt namn eller material Pyrithiamine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H19N4O.2BrH/c1-10-12(5-7-19)4-3-6-18(10)9-13-8-16-11(2)17-14(13)15;;/h3-4,6,8,19H,5,7,9H2,1-2H3,(H2,15,16,17);2*1H/q+1;;/p-1
Formel vikt 418.0004
Procent renhet >95%