Läs mer
Pyropheophorbide A Methyl Ester, TRC
Produktkod. 30413099
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30413099 50 mg
1 options
Produktkod. 30413099 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99886550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyropheophorbide A Methyl ester
CAS 6453-67-4
Molekylformel C34 H36 N4 O3
Kvantitet 50 mg
Synonym 3-Phorbinepropanoic acid','9-ethenyl-14-ethyl-4,8,13,18-tetramethyl-20-oxo-','methyl ester','(3S-trans)-, 3-Phorbinepropionic acid','14-ethyl-4,8,13,18-tetramethyl-20-oxo-9-vinyl-','methyl ester (8CI), Pyropheophorbide a methyl ester (6CI,7CI), MePPP, Methyl pyrophaeophorbide a, Methyl pyropheophorbide a, NSC 267052
InChI formel InChI=1S/C34H36N4O3/c1-8-20-16(3)24-13-26-18(5)22(10-11-31(40)41-7)33(37-26)23-12-30(39)32-19(6)27(38-34(23)32)15-29-21(9-2)17(4)25(36-29)14-28(20)35-24/h8,13-15,18,22,35,38H,1,9-12H2,2-7H3/b24-13-,25-14-,26-13-,27-15-,28-14-,29-15-,33-23-/t18-,22-/m0/s1
LEDER CCc1c(C)c2cc3[nH]c(cc4nc([C@@H](CCC(=O)OC)[C@@H]4C)c5CC(=O)C6=C(C)c(cc1n2)[nH]c56)c(C)c3C=C
Molekylvikt (g/mol) 548.67
Formel vikt 548.2787