missing translation for 'onlineSavingsMsg'
Läs mer

Pyrraline, TRC

Produktkod. 30012607
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
2.5 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012607 2.5 mg
30005643 5 mg
30011842 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30012607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9977802.5MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 74509-14-1
Molekylformel C12 H22 N2 O4
Molekylvikt (g/mol) 258.31
IUPAC-namn (2S)-2-amino-6-[2-formyl-5-(hydroxymethyl)pyrrolidin-1-yl]hexanoic acid
LEDER N[C@@H](CCCCN1C(CO)CCC1C=O)C(=O)O

Specifikationer

Kemiskt namn eller material Pyrraline
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C12H22N2O4/c13-11(12(17)18)3-1-2-6-14-9(7-15)4-5-10(14)8-16/h7,9-11,16H,1-6,8,13H2,(H,17,18)/t9?,10?,11-/m0/s1
Formel vikt 258.158
Procent renhet >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.