Läs mer
Pyruvic Acid-13C Sodium Salt, TRC
Produktkod. 30414658
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30414658 250 mg
1 options
Produktkod. 30414658 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP998896250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium Pyruvate-3-13C
CAS 124052-04-6
Molekylformel 13C C2 H3 O3 . Na
Kvantitet 250 mg
Synonym Propanoic-3-13C acid','2-oxo-','sodium salt (9CI), Sodium pyruvate-3-13C, Sodium [3-13C]pyruvate, Sodium pyruvate-13C, Sodium 2-oxopropanoate-3-13C, Sodium α-ketopropionate-γ-13C, Pyruvic acid sodium salt-13C, 2-Oxopropanoic-3-13C acid monosodium salt
InChI formel InChI=1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1;
LEDER [Na+].[13CH3]C(=O)C(=O)[O-]
IUPAC-namn sodium;2-oxo(3^{13}C)propanoate
Molekylvikt (g/mol) 111.037
Formel vikt 111.001