Läs mer
Pyruvic Acid-13C2, TRC
Produktkod. 30414128
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414128 1 mg
30424761 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414128 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9989071MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127350-53-2
Molekylformel C13C2H4O3
Molekylvikt (g/mol) 90.05
Synonym 2-oxo-Propanoic-1,2-13C2 acid
IUPAC-namn 2-oxo(2,3-^{13}C_{2})propanoic acid
LEDER [13CH3][13C](=O)C(=O)O
Specifikationer
Kemiskt namn eller material Pyruvic Acid-13C2
Kvantitet 1 mg
InChI formel InChI=1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)/i1+1,2+1
Formel vikt 90.0228