Läs mer
Pyruvic Acid-13C3 Sodium Salt, TRC
Produktkod. 30009854
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009854 25 mg
30008383 5 mg
30006257 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99890425MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142014-11-7
Molekylformel 13C3H3NaO3
Molekylvikt (g/mol) 113.02
Synonym 2-Oxopropanoic Acid13C3 Sodium Salt, Sodium Pyruvate13C3, Sodium α-Ketopropionate13C3
IUPAC-namn sodium;2-oxo(1,2,3-^{13}C_{3})propanoate
LEDER [13CH3][13C]([13C](O[Na])=O)=O
Specifikationer
Kemiskt namn eller material Pyruvic Acid-13C3 Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1,2+1,3+1;
Formel vikt 113.0081
Procent renhet >95%