Läs mer
QC6352, MedChemExpress
Produktkod. 16496657
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496657 5 mg
16480297 50 mg
16454966 100 mg
16431049 1 mg
16413948 10 mg
16412926 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16496657 Leverantör Medchem Express Leverantörsnummer HY104048
MedChemExpress QC6352 is an orally available, selective and potent KDM4C inhibitor with an IC50 of 35 nM.
QC6352 is an orally available, selective and potent KDM4C inhibitor with an IC50 of 35 nM.
Specifikationer
Kemiskt namn eller material QC6352
CAS 1851373-36-8
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C24H25N3O2
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 25 mg/mL (64.52 mM)
LEDER O=C(C1=C(NC[C@@H]2CCCC3=C2C=CC(N(C)C4=CC=CC=C4)=C3)C=NC=C1)O
Molekylvikt (g/mol) 387.47
Formel vikt 387.47
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death