missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 585 ITK™ Streptavidin Conjugate Kit
Qdot™ 585 ITK™ Streptavidin Conjugate Kit
Brand: Molecular Probes™ Q10011MP
Additional Details : Weight : 0.00450kg
Description
The Qdot™ 585 ITK™ Streptavidin Conjugate Kit provides the Qdot™ streptavidin conjugate as 250μL of a 2μM solution. Unlike the Qdot™ streptavidin conjugates, the Qdot™ ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo™ conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot™ incubation buffer.Specifications
Qdot 585 | |
Room Temperature | |
Conjugate Kit |
Store in refrigerator (2–8°C). | |
ITK™, Qdot™ |
For Research Use Only. Not for use in diagnostic procedures.