Läs mer
Tocris Bioscience™ QO 58
Kv7 channel opener
Brand: Tocris Bioscience™ 5083/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
QO 58 is a Kv7 channel opener (EC50 values are 0.6, 1.0, 5.2 and 7.0 μM for Kv7.4, Kv7.2, Kv7.3/7.5 and Kv7.1, respectively). Increases the pain threshold of neuropathic pain in a sciatic nerve CCI in vivo model.Specifikationer
| 5-(2,6-Dichloro-5-fluoro-3-pyridinyl)-3-phenyl-2-(trifluoromethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one | |
| 1259536-62-3 | |
| 10 mg |