missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QPRT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25174-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
QPRT gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer′s disease, and Huntington′s disease. Alternative splicing results in multiple transcript variants.Specifications
QPRT | |
Polyclonal | |
Unconjugated | |
QPRT | |
QAPRTase, QPRT, QPRTase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23475 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15274 | |
QPRT | |
QPRT Fusion Protein Ag17680 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |