Learn More
Hellma™ QS Quartz Suprasil High Precision Ultra Macro Fluorescence Cells With Polyethylene Stopper
Brand: Hellma™ 105-250-15-40
Additional Details : Weight : 0.80000kg
Specifications
Ultra-macro Fluorescence Cell | |
Quartz Suprasil | |
45 mm |
100 μL | |
10 x 2 mm | |
12.5 mm |