Learn More
R&D Systems™ Quantikine Immunoassay Control 1035 for Human ErbB3/Her3
Product Code. 30000911
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
30000911 1 Each Each
1 options
Product Code. 30000911 Supplier R&D Systems™ Supplier No. QC275
  • ONLINE EXCLUSIVE PRICE
    2282.89 SEK
    2013.90 SEK / Paket med 1
    Save 268.99 SEK
    11% Av
 Restricted Item
Beräknad leverans: 26-11-2026
For use with catalog number DERB30
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Gene ID (Entrez) 265
Product Line R&D Systems
Quantity 1 Each
Source Human
Target Enzyme ErbB3/Her3