Learn More
Fisherbrand™ Quantitative Grade Filter Paper Circles
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
POA
Specifications
Format Disc
Material Alpha Cotton Cellulose
Quantity 100/box
View More Specs
Description
Description
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
  • Made from high-quality alpha cotton cellulose, washed in hydrochloric acid and neutralized in an ultrapure water rinse
  • Leave no more than 0.01% ash when burned
  • Limited wet strength, suitable for gravity or low-suction filtration; should be used with care to avoid rupturing
Order Info
Packed 100 circles to a box and shrink-wrapped for purity and cleanliness.
Compliance
All papers listed in chart meet ASTM™ requirements for “ash content per circle.”
Specifications
Specifications
Disc
Alpha Cotton Cellulose
100/box