Läs mer
Applied Biosystems™ QuantStudio™ 3D Digital PCR 20K Chip Kit v2
Produktkod. 16205090 Handla allt Applied Biosystems Produkter
Produktkod. 16205090
Brand: Applied Biosystems™ A26316
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Key component of the QuantStudio™ 3D Digital PCR System.
Manufactured from a silicon substrate and etched with 20,000 consistently-sized nanoscale reaction wells, the chip enables precise sample loading with an uncomplicated workflow. The QuantStudio 3D Digital PCR 20K Chip Kit v2 enables improved UV adhesive-free sealing. It requires use of the QuantStudio 3D™ Digital PCR Master Mix v2 and is fully-compatible with all QuantStudio 3D™ Digital PCR Instrument hardware and software. The QuantStudio 3D Digital PCR 20K Chip Kit v2 is designed to deliver accurate, sensitive, and precise digital PCR data on QuantStudio 3D Digital PCR instruments when combined with associated reagents and QuantStudio™ 3D Analysis Suite™ software.
For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring • 12 QuantStudio 3D Digital PCR 20K Chips v2
• 12 QuantStudio 3D Digital PCR Chip Lids v2
• 12 QuantStudio 3D Digital PCR Sample Loading Blades
• 3 syringes Immersion Fluid
• 3 Immersion Fluid Tips
Store at room temperature.
För användning med (utrustning) QuantStudio™ 3D
Produktlinje QuantStudio
Produkttyp 3D Digital PCR Chip Kit
För användning med (applikation) Real Time PCR (qPCR)
Frakt skick Room Temperature
Beskrivning Version 2
Kvantitet 12 chips