Learn More
Hellma™ Quartz Suprasil 4 Faces Cuveet
Brand: Hellma™ 117-104-10-40
Additional Details : Weight : 0.80000kg
Specifications
Cuvette | |
Quartz Suprasil | |
200 to 2500 nm |
4 Faces | |
2Pack |