Läs mer
Quercitrin, TRC
Produktkod. 30137973
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137973 50 mg
30012213 10 mg
30004951 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ50952050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 522-12-3
Molekylformel C21 H20 O11
Molekylvikt (g/mol) 448.38
Synonym 4H-1-Benzopyran-4-one, 3-[(6-deoxy-α-L-mannopyranosyl)oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-, Quercitrin (7CI,8CI), 3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone 3-α-L-rhamnoside, 3-O-α-L-Rhamnopyranosylquercetin, 5,7,3',4'-Tetrahydroxyflavone 3-O-α-L-rhamnoside, C.I. 75720, NSC 9221, Quercetin 3-O-rhamnoside, Quercetin 3-O-α-L-rhamnopyranoside, Quercetin 3-O-α-L-rhamnoside, Quercetin 3-O-α-rhamnopyranoside, Quercetin 3-O-α-rhamnoside, Quercetin 3-rhamnoside, Quercetin 3-α-L-rhamnoside, Quercetin 3-α-rhamnoside, Quercetin rhamnoside, Quercimelin, Quercitroside, WA 17779
IUPAC-namn 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
LEDER C[C@@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Quercitrin
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C21H20O11/c1-7-15(26)17(28)18(29)21(30-7)32-20-16(27)14-12(25)5-9(22)6-13(14)31-19(20)8-2-3-10(23)11(24)4-8/h2-7,15,17-18,21-26,28-29H,1H3/t7-,15-,17+,18+,21-/m0/s1
Formel vikt 448.1006
Procent renhet >95%