Läs mer
Quetiapine Carboxylate Impurity, TRC
Produktkod. 30413411
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413411 250 mg
30418663 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ509980250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1011758-00-1
Molekylformel C22 H25 N3 O4 S
Molekylvikt (g/mol) 427.52
Synonym 1-Piperazinecarboxylic acid, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, 2-(2-hydroxyethoxy)ethyl ester, 2-(2-Hydroxyethoxy)ethyl 4-Dibenzo[b,f][1,4]thiazepin-11-ylpiperazine-1-carboxylate
IUPAC-namn 2-(2-hydroxyethoxy)ethyl 4-benzo[b][1,4]benzothiazepin-6-ylpiperazine-1-carboxylate
LEDER OCCOCCOC(=O)N1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24
Specifikationer
Kemiskt namn eller material 2-(2-Hydroxyethoxy)ethyl 4-Dibenzo[b,f][1,4]thiazepin-11-ylpiperazine-1-carboxylate
Kvantitet 250 mg
InChI formel InChI=1S/C22H25N3O4S/c26-13-14-28-15-16-29-22(27)25-11-9-24(10-12-25)21-17-5-1-3-7-19(17)30-20-8-4-2-6-18(20)23-21/h1-8,26H,9-16H2
Formel vikt 427.1566