Läs mer
Quetiapine Dimer Impurity, TRC
Produktkod. 30414372
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30414372 25 mg
1 options
Produktkod. 30414372 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ50996025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Quetiapine Dimer Impurity
CAS 945668-94-0
Molekylformel C30 H24 N4 S2
Kvantitet 25 mg
Synonym Dibenzo[b,f][1,4]thiazepine','11,11'-(1,4-piperazinediyl)bis-, 11,11'-(1,4-Piperazinediyl)bis-dibenzo[b,f][1,4]thiazepine, 11,11'-(1,4-Piperazinediyl)bis[dibenzo[b,f][1,4]thiazepine], Quetiapine Fumarate Imp. D (EP), Quetiapine dimer impurity, Bis(dibenzothiazepinyl) piperazine, 1,4-Bis(dibenzo[b,f][1,4]thiazepin-11-yl)piperazine
InChI formel InChI=1S/C30H24N4S2/c1-5-13-25-21(9-1)29(31-23-11-3-7-15-27(23)35-25)33-17-19-34(20-18-33)30-22-10-2-6-14-26(22)36-28-16-8-4-12-24(28)32-30/h1-16H,17-20H2
LEDER C1CN(CCN1C2=Nc3ccccc3Sc4ccccc24)C5=Nc6ccccc6Sc7ccccc57
IUPAC-namn 6-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)benzo[b][1,4]benzothiazepine
Molekylvikt (g/mol) 504.67
Formel vikt 504.1442