Läs mer
Quetiapine Impurity-N, TRC
Produktkod. 17982505
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982505 100 mg
17974465 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ510015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1800291-86-4
Molekylformel C29 H41 N5 O3 S
Molekylvikt (g/mol) 539.73
Synonym 2-[2-[4-[2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethyl]-1-piperazinyl]ethoxy]ethanol, Quetiapine Fumarate Imp. N (EP)
IUPAC-namn 2-[2-[4-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethyl]piperazin-1-yl]ethoxy]ethanol
LEDER OCCOCCN1CCN(CCOCCN2CCN(CC2)C3=Nc4ccccc4Sc5ccccc35)CC1
Specifikationer
Kemiskt namn eller material 2-[2-[4-[2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethyl]-1-piperazinyl]ethoxy]ethanol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C29H41N5O3S/c35-20-24-37-23-19-32-11-9-31(10-12-32)17-21-36-22-18-33-13-15-34(16-14-33)29-25-5-1-3-7-27(25)38-28-8-4-2-6-26(28)30-29/h1-8,35H,9-24H2
Formel vikt 539.293
Procent renhet >95%