Learn More
Quickfit™ Allihn Condenser with Ground Cone
Product Code. 11371845
Change view
Click to view available options
Standard Taper Joint No:
19/26
24/29
29/32
34/35
40/38
50/42
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
11371845 50/42 Each
11338074 19/26 Each
11348074 24/29 Each
12030073 29/32 Each
11351845 34/35 Each
11361845 40/38 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11371845 Supplier Quickfit Supplier No. CX7/08/SC
  • ONLINE EXCLUSIVE PRICE
    2565.00 SEK
    2115.88 SEK / Paket med 1
    Save 449.12 SEK
    17% Av
View all promotions
Beräknad leverans: 18-02-2027
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
For Use With (Application) Ideal for reflux work
Length (Metric) Jacket 365 mm
Length (Metric) 365 mm
Standard Taper Joint No 50/42
Material Borosilicate Glass
Type Allihn Condenser