Läs mer
Quickfit™ Borosilicate Glass Ether Condenser
Standard konisk led nr:
19/26
Förpackningsstorlek:
Paket med 2
Beskrivning
- Highly efficient coil condenser suitable for condensing very volatile liquids
- Inner coil with large surface area
- Outer water jacket to avoid vapour creep around the coil
- Coil and jacket are independent and can be fed from separate water supplies for maximum efficiency
- With detachable plastic screwthread connectors for the easy and safe attachment of flexible tubing
Specifikationer
Specifikationer
| För användning med (applikation) | Suitable for condensing very volatile liquids |
| Standard konisk led nr | 19/26 |
| Material | Borosilicate Glass |
| Typ | Ether Condenser |