Learn More
Quickfit™ Borosilicate Glass Light Entrainer Receiver
Determine the water content in a solution with the Dean-Stark pattern apparatus. QuickFit™ borosilicate glass light entrainer receiver.
Brand: Quickfit WE6/23/7
1282.50 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Complies with BS 756
- Dean and Stark pattern receivers for the determination of the water content in petroleum products
- For use with solvent entrainers which are lighter than water
- Borosilicate glass with graduated tube
Specifications
For use with solvent entrainers which are lighter than water | |
Borosilicate Glass | |
BS 756 | |
Light entrainer |
Light Entrainer Receiver | |
7.5 mL | |
7.5 mL | |
Cone: 24/29, Socket: 19/26 |