Learn More
Quickfit™ Borosilicate Glass Round Bottom Flask with Septum Side Neck
Designed for semi-micro and micro scale preparations.
2520.00 SEK - 3750.00 SEK
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Designed for semi-micro and micro scale preparations |
Neck Style | Septum side neck |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15577850
|
Quickfit
FR10/1S/SEP |
10 mL |
3750.00 SEK
Pack of 5 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
15527870
|
Quickfit
FR25/1S/SEP |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- One vertical centrer neck having precision-ground Quickfit™ socket to accept Quickfit™ cone
- One angled side neck designed to take a rubber septum seal to allow small quantities of liquid to be handled easily and safely using a syringe
Specifications
Borosilicate Glass | |
Designed for semi-micro and micro scale preparations | |
Septum side neck | |
Round |