Learn More
Quickfit™ Borosilicate Glass Vacuum Filter Flask with Glass Sintered Disc
Brand:  Quickfit VB40/04M
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 15578000
  • 2074.00 SEK / Pack of 5
Estimated Shipment
28-12-2020
Add to basket
Description
Description
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
Specifications
Specifications
ISO 3585, DIN 12217, ASTM E-438
Borosilicate Glass
52 mm
Buchner Vacuum Filtration Funnel
40 mm
80 mL
For filtering solutions that can be filtered by filter papers which are not resistant to certain acids and bases
Quickfit™ ground Glass cone
24/29
2