Learn More
Quickfit™ Chromatography Column with Ground Cone
Manufactured from borosilicate glass. Quickfit™ Chromatography Column with Ground Cone includes a 0 porosity chemically resistant glass sintered disc. Upper socket can be connected to a solvent reservoir. Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow.
Brand:  Quickfit CR32/40
Additional Details : Weight : 0.23000kg
 View more versions of this product
Product Code. 15557770
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • With glass sintered disc - porosity 0 to support column packing
  • Upper socket can be connected to a solvent reservoir
  • Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow
  • Manufactured from borosilicate glass
Specifications
Specifications
18 mm
1 Pack
Pyrex™ Borosilicate Glass
Glass sintered disc
Chromatography Column
515 mm
19/26 Cone, 24/29 Socket