Learn More
Quickfit™ Glass Stirrer Shaft
Designed for use with SVL™ stirrer heads and stirrer bearings
Brand:  SVL 588-34
Additional Details : Weight : 0.11000kg
 View more versions of this product
Product Code. 15507740
  • 6261.02 SEK / Pack of 5
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Manufactured from borosilicate glass
  • Lower end with internal screwthread to accept choice of stirrer heads
  • Upper section of shaft is ground to fit choice of stirrer bearings
Specifications
Specifications
1PK
Length: 490mm
SVL™ stirrer heads and stirrer bearings
Stirrer shaft
Videos