Learn More
Quickfit™ Metal Clamps for Flat Flange Joints
Offered in two versions: fork design and spring wire design. Quickfit™ Metal Clamps for Flat Flange Joints are designed to secure corresponding Quickfit™ glassware with flat flange joints.
Brand:  Quickfit JC51
Additional Details : Weight : 0.26000kg
 View more versions of this product
Product Code. 12049082
  • 801.11 SEK / Pack of 2
Estimated Shipment: 24-08-2021
Add to basket
Description
Description
Offered in two versions: fork design and spring wire design. Quickfit™ Metal Clamps for Flat Flange Joints are designed to secure corresponding Quickfit™ glassware with flat flange joints.
Two distinct styles to meet joint needs
  • Fork design: spring loaded to ensure balanced positive pressure to both joint surfaces, finished with corrosion-resistant nickel, suitable for use with spherical joints
  • Spring wire design: four touch points for stability, secure swing clamp, stainless steel
Specifications
Specifications
Spring wire design
Joint Clamp
Stainless Steel
Glassware with flat flange joints