Learn More
Quickfit™ Plain Short Stem Adapter
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
Marque: Quickfit RA1/04
1676.75 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Informations supplémentaires : Weight : 0.04000kg
Description
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
- With short or long stem
- Useful for feeding distillate from condensers into receiving vessels
Spécification
Receiver Adapter | |
2.55 in. | |
Feeding distillate from condensers into receiving vessels |
Borosilicate Glass | |
65 mm | |
29/32 |