missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Three-Vertical-Neck Round-Bottom Flask

Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.

Brand:  Quickfit FR500/3S/22P

4897.25 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15507900

  • 5155.00 SEK / Pack of 5
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

  • Fitted with three precision-ground sockets
    • One vertical center neck
    • Two vertical side necks
  • Ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
Specifications

Specifications

500 mL
150 mm
24/29 (Centre), 19/26 (side)
Borosilicate Glass
3
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers