Läs mer
Contec Quicktask Non-Magnetic Mop Frame, Stainless Steel
Produktkod. 31723042 Handla allt Contec Produkter
Change view
Autoklaveringsbar:
Autoclavable
Produktkod. Autoklaveringsbar
31723042 Autoclavable
1 options
Produktkod. 31723042 Leverantör Contec Leverantörsnummer 6208
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Flat hinged mop head frames are durable, lightweight and exclusively designed for the QuickTask™ Mop System.
  • Available in stainless steel or blue polypropylene
  • Autoclavable
  • Compatible with most common disinfectant solutions and solvents
  • The QuickTask™ system consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly
  • The QuickTask™ system is ideally suited for cleaning floors, including perforated cleanroom floors and grates
TRUSTED_SUSTAINABILITY
Specifikationer
Produkttyp 1 Piece Lay Flat Frame
Sterilitet Non-sterile
Autoklaveringsbar Autoclavable
Disponibel Reusable
Färgkodad No
Material Stainless Steel