Läs mer
Quiflapon sodium, MedChemExpress
Produktkod. 16497856
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497856 100 mg -
16430558 5 mg 5mg
16418098 10 mg -
16404017 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16497856 Leverantör Medchem Express Leverantörsnummer HY50714
MedChemExpress Quiflapon sodium (MK-591 sodium) is a selective and specific 5-Lipoxygenase-activating protein (FLAP) inhibitor. Quiflapon sodium is an orally active Leukotriene biosynthesis inhibitor. Induces apoptosis.
Quiflapon sodium (MK-591 sodium) is a selective and specific 5-Lipoxygenase-activating protein (FLAP) inhibitor. Quiflapon sodium is an orally active Leukotriene biosynthesis inhibitor. Induces apoptosis.
Specifikationer
Kemiskt namn eller material Quiflapon sodium
CAS 147030-01-1
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C34H34ClN2NaO3S
Kvantitet 100 mg
Synonym MK-591 sodium
Löslighetsinformation DMSO : 50 mg/mL (82.08 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER ClC(C=C1)=CC=C1CN2C(CC(C)(C(O[Na])=O)C)=C(SC(C)(C)C)C3=C2C=CC(OCC4=NC5=CC=CC=C5C=C4)=C3
Molekylvikt (g/mol) 609.15
Formel vikt 609.15
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation