Läs mer
Quillaic Acid, TRC
Produktkod. 30135983
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135983 100 mg
30012832 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ550500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 631-01-6
Molekylformel C30H46O5
Molekylvikt (g/mol) 486.68
Synonym 3β,16α-Dihydroxy-23-oxo-olean-12-en-28-oic Acid, Quillaja Sapogenin
IUPAC-namn (4aR,5R,6aR,6aS,6bR,8aR,9S,10S,12aR,14bS)-9-formyl-5,10-dihydroxy-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
LEDER [Na+].[Na+].Nc1cc(S(=O)(=O)[O-])c(cc1)CCc2c(cc(N)cc2)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Quillaic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C30H46O5/c1-25(2)13-14-30(24(34)35)19(15-25)18-7-8-21-26(3)11-10-22(32)27(4,17-31)20(26)9-12-28(21,5)29(18,6)16-23(30)33/h7,17,19-23,32-33H,8-16H2,1-6H3,(H,34,35)/t19-,20+,21+,22-,23+,26-,27-,28+,29+,30+/m0/s1
Formel vikt 486.3345