Läs mer
Tocris Bioscience™ Quin C1
Potent and selective FPR2 agonist
Brand: Tocris Bioscience™ 5583/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Quin C1 is a potent and selective FPR2 agonist (EC50 = 15 nM). Induces Ca2+ mobilization in FPR2 but not FPR1-transfected mast cells. Induces neutrophil chemotaxis and degranulation in vitro. Reduces neutrophil and lymphocyte counts in bronchoalveolar lavage fluid in a mouse lung injury model. Anti-inflammatory.Specifikationer
| 4-Butoxy-N-[1,4-dihydro-2-(4-methoxyphenyl)-4-oxo-3(2H)-quinazolinyl]benzamide | |
| 786706-21-6 | |
| 10 mg |