Läs mer
Quinacrine Dihydrochloride, TRC
Produktkod. 30421968
Change view
Kvantitet:
50 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421968 10 g
30423912 500 mg
30411283 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421968 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ55061010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69-05-6
Molekylformel C23 H30 Cl N3 O . 2 Cl H
Molekylvikt (g/mol) 472.88
Synonym Mepacrine Hydrochloride, 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, hydrochloride (1:2), 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, dihydrochloride (9CI), Acridine, 6-chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxy-, dihydrochloride (8CI), 2-Chloro-5-(ω-diethylamino-α-methylbutylamino)-7-methoxyacridine dihydrochloride, 2-Methoxy-6-chloro-9-(4-diethylamino-1-methylbutylamino)acridine dihydrochloride, 3-Chloro-7-methoxy-9-(1-methyl-4-diethylaminobutylamino)acridine dihydrochloride, 3-Chloro-9-(4'-diethylamino-1'-methylbutylamino)-7-methoxyacridine dihydrochloride, 6-Chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxyacridine dihydrochloride, 866RP, Acrichin dihydrochloride, Atabrine hydrochloride, Atebrin, Atebrine, Atebrine hydrochloride, Chemiochin, Chinacrin hydrochloride, Crinodora, Erion, Malaricida, Mecryl, Mepacrine dihydrochloride, Mepacrine hydrochloride, Methoquine, Metochin, Metoquin, Metoquine, NSC 14229, Palacrin, Palusan, Pentilen, Quinacrine dihydrochloride, Quinacrine hydrochloride, SN 390
IUPAC-namn 4-N-(6-chloro-2-methoxyacridin-9-yl)-1-N,1-N-diethylpentane-1,4-diamine;dihydrochloride
LEDER Cl.Cl.CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13
Specifikationer
Kemiskt namn eller material Mepacrine Hydrochloride
Kvantitet 10 g
InChI formel InChI=1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H
Formel vikt 471.1611