missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Quinaldine Red, dye content, 95%
Click to view available options
Quantity:
5 g
25 g
Description
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).
Solubility
Partly soluble in water, alcohol, and ethanol.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).
Solubility
Partly soluble in water, alcohol, and ethanol.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
Color | Black to Green |
Quantity | 5 g |
Beilstein | 3798055 |
Sensitivity | Light sensitive |
Merck Index | 14,8049 |
Solubility Information | Partly soluble in water,alcohol,and ethanol. |
Formula Weight | 430.33 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | Quinaldine Red, Dye content |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'