Learn More
Quinapril hydrochloride, 98%
Product Code. 15494399
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494399 100 mg 100mg
15474399 1 g 1g
15484399 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15494399 Supplier Thermo Scientific Alfa Aesar Supplier No. J61913.MC
 Restricted Item
I lager
Add to basket
Angiotensin-converting enzyme inhibitor | CAS: 82586-55-8 | C25H31ClN2O5 | 474.982 g/mol
Angiotensin-converting enzyme inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82586-55-8
Molecular Formula C25H31ClN2O5
Molecular Weight (g/mol) 474.982
MDL Number MFCD00889215
InChI Key IBBLRJGOOANPTQ-JKVLGAQCSA-N
Synonym quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
PubChem CID 54891
ChEBI CHEBI:8714
IUPAC Name (3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl
Specifications
Melting Point 120°C to 130°C
Color White
Quantity 100 mg
Merck Index 14,8051
Formula Weight 474.99
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Quinapril hydrochloride