Läs mer
Quinestrol, TRC
Produktkod. 30425911
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30425911 100 mg
1 options
Produktkod. 30425911 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ670403100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Quinestrol
CAS 152-43-2
Molekylformel C25 H32 O2
Kvantitet 100 mg
Synonym 19-Nor-17α-pregna-1,3,5(10)-trien-20-yn-17-ol','3-(cyclopentyloxy)- (6CI,7CI,8CI), (17α)-3-(Cyclopentyloxy)-19-norpregna-1,3,5(10)-trien-20-yn-17-ol, 17-Ethynylestradiol cyclopentyl ether, 17α-Ethynylestradiol 3-cyclopentyl ether, 3-(Cyclopentyloxy)-19-nor-17α-pregna-1,3,5(10)-trien-20-yn-17-ol, 3-Cyclopentyloxy-17α-ethynyl-1,3,5(10)-estratrien-17-ol, 3-Cyclopentyloxy-17α-ethynylestra-1,3,5(10)-trien-17β-ol, EECPE, Eston, Estrovis, Estrovis 4000, Estrovister, Ethynylestradiol-3-cyclopentyl ether, Plestrovis, Qui-Lea, Quinestrol, W 3566
InChI formel InChI=1S/C25H32O2/c1-3-25(26)15-13-23-22-10-8-17-16-19(27-18-6-4-5-7-18)9-11-20(17)21(22)12-14-24(23,25)2/h1,9,11,16,18,21-23,26H,4-8,10,12-15H2,2H3/t21-,22-,23+,24+,25+/m1/s1
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(OC5CCCC5)ccc34)[C@@H]1CC[C@@]2(O)C#C
IUPAC-namn (8R,9S,13S,14S,17R)-3-cyclopentyloxy-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-ol
Molekylvikt (g/mol) 364.52
Formel vikt 364.2402