Läs mer
Quinidine Methiodide, TRC
Produktkod. 30134457
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134457 1 g
17981715 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134457 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ6850201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42982-87-6
Molekylformel C21H27IN2O2
Molekylvikt (g/mol) 466.36
Synonym Quinidine α-Methiodide, (9S)-9-Hydroxy-6'-methoxy-1-methylcinchonanium Iodide
IUPAC-namn (S)-[(1S,2R,4S,5R)-5-ethenyl-1-methyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;iodide
LEDER [H][C@@]1(C=C)[C+]2C[C@@]([C@@H](O)C3=CC=NC4=CC=C(OC)C=C43)([H])[N@@](C1)(C)CC2.[I-]
Specifikationer
Kemiskt namn eller material Quinidine Methiodide
Kvantitet 1 g
InChI formel InChI=1S/C21H27N2O2.HI/c1-4-14-13-23(2)10-8-15(14)11-20(23)21(24)17-7-9-22-19-6-5-16(25-3)12-18(17)19;/h4-7,9,12,14-15,20-21,24H,1,8,10-11,13H2,2-3H3;1H/q+1;/p-1/t14-,15-,20+,21-,23-;/m0./s1
Formel vikt 466.1117