Läs mer
Quinine Hydrochloride, TRC
Produktkod. 30426305
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30426305 1 g
1 options
Produktkod. 30426305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ6940201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Quinine hydrochloride
CAS 130-89-2
Molekylformel C20 H24 N2 O2 . Cl H
Kvantitet 1 g
Synonym Cinchonan-9-ol','6'-methoxy-','monohydrochloride','(8α,9R)- (9CI), Quinine','monohydrochloride (8CI), Chinimetten, Chinine hydrochloride, Quinine hydrochloride, Quinine muriate, (R)-[(2S,4S,5R)-5-Ethenyl-1-azabicyclo[2.2.2]oct-2-yl]-(6-methoxyquinolin-4-yl)methanol hydrochloride
InChI formel InChI=1S/C20H24N2O2.ClH/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H/t13-,14-,19-,20+;/m0./s1
LEDER Cl.COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1
IUPAC-namn (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
Molekylvikt (g/mol) 360.88
Formel vikt 360.1605