Learn More
Quinoline, 96%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
Solubility
More soluble in hot than cold water. Miscible with alcohol, ether
Notes
Dissolves sulfur, As{2}O{3}, phosphorus
Specifications
Melting Point | -15°C |
Density | 1.09 |
pH | 7.3 |
Boiling Point | 237°C |
Flash Point | 101°C (214°F) |
Refractive Index | 1.626 |
Quantity | 100 g |
UN Number | UN2656 |
Beilstein | 107477 |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
RUO – Research Use Only