Läs mer
Quinoline Yellow, TRC
Produktkod. 30136682
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136682 5 g
30133836 10 g
17942615 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136682 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ7010405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 8003-22-3
Molekylformel C18H11NO2
Molekylvikt (g/mol) 269.3
Synonym C.I. Solvent Yellow 33, 1,3-Isobenzofurandione, reaction products with dimethylquinoline and methylquinoline, Arlosol Yellow S, C.I. 47000, Chinoline Yellow D Sol.in Spirits, Chinoline Yellow ZSS, D & C Yellow 11, D and C Yellow No. 11, D&C Yellow No. 11, Japan Yellow 204, Japan Yellow No. 204, Kayaset Yellow 2G, Nitro Fast Yellow SL, Oil Yellow SIS, Oplas Yellow 136, Petrol Yellow C, Polysolve Yellow 33G, Quinoline Yellow A Spirit Soluble, Quinoline Yellow Base, Quinoline Yellow SS, Quinoline Yellow Spirit Soluble, Solvent Yellow 33, Waxoline Yellow T, Yellow 11, C.I. 47000
IUPAC-namn 2-(6-methyl-1H-quinolin-2-ylidene)indene-1,3-dione
LEDER Cc1ccc2NC(=C3C(=O)c4ccccc4C3=O)C=Cc2c1
Specifikationer
Kemiskt namn eller material Solvent Yellow 33
Kvantitet 5 g
InChI formel InChI=1S/C19H13NO2/c1-11-6-8-15-12(10-11)7-9-16(20-15)17-18(21)13-4-2-3-5-14(13)19(17)22/h2-10,20H,1H3
Formel vikt 287.0946