Läs mer
Quinotoxine Hydrochloride, TRC
Produktkod. 30136567
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136567 500 mg
30009630 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136567 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ753500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52211-63-9
Molekylformel C20H24N2O2 . HCl
Molekylvikt (g/mol) 360.9
Synonym 3-[(3R,4R)-3-Ethenyl-4-piperidinyl]-1-(6-methoxy-4-quinolinyl)-1-propanone Hydrochloride, (3R-cis)-3-(3-Ethenyl-4-piperidinyl)-1-(6-methoxy-4-quinolinyl)-1-propanone Hydrochloride, Quinicine Hydrochloride, (3R,4R)-4-[3-Oxo-3-(6-methoxyquinolin-4-yl)propyl]-3-vinylpiperidine Hydrochloride, Chinicine Hydrochloride, Chinotoxin, Permiran, Viquidil Hydrochloride;
IUPAC-namn 3-[(3R,4R)-3-ethenylpiperidin-4-yl]-1-(6-methoxyquinolin-4-yl)propan-1-one;hydrochloride
LEDER Cl.COc1ccc2nccc(C(=O)CC[C@@H]3CCNC[C@@H]3C=C)c2c1
Specifikationer
Kemiskt namn eller material Quinotoxine Hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C20H24N2O2.ClH/c1-3-14-13-21-10-8-15(14)4-7-20(23)17-9-11-22-19-6-5-16(24-2)12-18(17)19;/h3,5-6,9,11-12,14-15,21H,1,4,7-8,10,13H2,2H3;1H/t14-,15+;/m0./s1
Formel vikt 360.1605