Läs mer
Quinupramine, TRC
Produktkod. 30010458
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010458 25 mg
30007637 50 mg
30007836 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010458 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ79670025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31721-17-2
Molekylformel C21 H24 N2
Molekylvikt (g/mol) 304.43
IUPAC-namn 11-(1-azabicyclo[2.2.2]octan-3-yl)-5,6-dihydrobenzo[b][1]benzazepine
LEDER C1CN2CCC1C(C2)N3c4ccccc4CCc5ccccc35
Specifikationer
Kemiskt namn eller material Quinupramine
Kvantitet 25 mg
InChI formel InChI=1S/C21H24N2/c1-3-7-19-16(5-1)9-10-17-6-2-4-8-20(17)23(19)21-15-22-13-11-18(21)12-14-22/h1-8,18,21H,9-15H2
Formel vikt 304.1939