missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ Quipazine dimaleate
GREENER_CHOICE

Produktkod. 30001109
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001109 Supplier Tocris Bioscience™ Supplier No. 0629/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT3 agonist

Selective 5-HT3 receptor agonist. Also displays antagonist activity at peripheral 5-HT3 receptors. [3H]-Quipazine labels 5-HT3 sites in the cortical membranes.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Quipazine dimaleate
CAS 150323-78-7
Kvantitet 100 mg
Molekylformel C21H23N3O8
Formel vikt 445.43
Renhet >99%
Molekylvikt (g/mol) 445.428
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.