missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), TRC
Specifications
Specifications
| Kemiskt namn eller material | Quizalofop-ethylD3(3,3,3-D3) |
| CAS | 1398065-84-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C19 2H3 H14 Cl N2 O4 |
| Kvantitet | 1 mg |
| Synonym | Quizalofop-ethyl D3 (3,3,3 D3), Quizalofop Ethyl D3 (methyl D3), Propanoic-3,3,3-d3 acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-','ethyl ester (ACI), Ethyl 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]propanoate-3,3,3-d3 (ACI), Quizalofop-p-ethyl-d3 |
| InChI formel | InChI=1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3/i2D3 |
| LEDER | [2H]C([2H])([2H])C(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC |
| IUPAC-namn | ethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]-3,3,3-trideuteriopropanoate |
| Molekylvikt (g/mol) | 375.82 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?