Läs mer
Quizalofop-ethyl, TRC
Produktkod. 30414814
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30414814 5 mg
1 options
Produktkod. 30414814 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ8261085MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Quizalofop-ethyl
CAS 76578-14-8
Molekylformel C19 H17 Cl N2 O4
Kvantitet 5 mg
Synonym Quizalofop Ethyl, Propanoic acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-','ethyl ester (9CI','ACI), Assure, DPX 6202, DPX-Y 6202, DPX-Y 6202-3, Ethyl 2-[4-(6-chloro-2-quinoxalyloxy)phenoxy]propionate, Miura, NC 302, NCI 96683, Quinofop-ethyl, Quizalofop-ethyl, Targa, Xylofop-ethyl
InChI formel InChI=1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3
LEDER CCOC(=O)C(C)Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1
IUPAC-namn ethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
Molekylvikt (g/mol) 372.8
Formel vikt 372.0877