Läs mer
Quizalofop-p-tefuryl, TRC
Produktkod. 30011728
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011728 10 mg
30008106 50 mg
30009080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011728 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ82611810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119738-06-6
Molekylformel C22 H21 Cl N2 O5
Molekylvikt (g/mol) 428.8655
Synonym Propanoic acid, 2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-, (tetrahydro-2-furanyl)methyl ester, Quizalofop-tefuryl, UBI 1956, (±) tetrahydrofurfuryl (R)-2-[4-(6-chloroquinoxalin-2-yloxy)phenyloxy]propionate
IUPAC-namn oxolan-2-ylmethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
LEDER CC(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC4CCCO4
Specifikationer
Kemiskt namn eller material Quizalofop-p-tefuryl
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H21ClN2O5/c1-14(22(26)28-13-18-3-2-10-27-18)29-16-5-7-17(8-6-16)30-21-12-24-20-11-15(23)4-9-19(20)25-21/h4-9,11-12,14,18H,2-3,10,13H2,1H3
Formel vikt 428.1139
Procent renhet >95%